Learn More
CPC Scientific H-Tyr-Gly-Gly-Phe-Leu-Arg-Arg-Ile-Arg-Pro-Lys-Leu-Lys-Trp-Asp-Asn-Gln-OH (trifluoroacetate salt) 0.5MG
SDP
Supplier:  CPC Scientific DYNP001A
Encompass
Prodynorphin (209-225). Opioid receptor agonist.ONE-LETTER SEQUENCE: YGGFLRRIRPKLKWDNQMOLECULAR FORMULA: C99H155N31O23MOLECULAR WEIGHT:2147.52STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [80448-90-4], SYNONYMS: Dynorphin A, Prodynorphin (209-225) (porcine), Prodynorphin (207-223) (human)RESEARCH AREA: PainREFERENCES: A. Goldstein et al., PNAS, 78, 7219 (1981); S. Tachibana et al., Nature, 295, 339 (1982)
Catalog No. 50-210-9201