Learn More
Bachem Dynorphin A (1-8) Acetate
SDP
Supplier:  Bachem 4005845.0005
Encompass_Preferred
Dynorphin A (1-8) Acetate, H-Tyr-Gly-Gly-Phe-Leu-Arg-Arg-Ile-OH, 75790-53-3, C46H72N14O10, Mr 981.17, 5mg. Dynorphin A (1-8), porcine.
Catalog No. 50-259-637