Learn More
Bachem Dynorphin A Trifluoroacetate
SDP
Supplier:  Bachem 4007832.0001
Encompass_Preferred
Dynorphin A Trifluoroacetate, H-Tyr-Gly-Gly-Phe-Leu-Arg-Arg-Ile-Arg-Pro-Lys-Leu-Lys-Trp-Asp-Asn-Gln-OH, 80448-90-4, C99H155N31O23, Mr 2147.52, 1mg. Prodynorphin (209-225) (porcine), Prodynorphin (207-223) (human), Dynorphin A (1-17). Endogenous kappa-receptor antagonist.
Catalog No. 50-259-634