Learn More
CPC Scientific H-Tyr-Gly-Gly-Phe-Leu-Arg-Arg-Ile-Arg-Pro-Lys-Leu-Lys-Trp-Asp-Asn-Gln-NH2 (trifluoroacetate salt) 0.5MG
SDP
Supplier:  CPC Scientific DYNP004A
Encompass
ONE-LETTER SEQUENCE: YGGFLRRIRPKLKWDNQ-NH2MOLECULAR FORMULA: C99H156N32O22MOLECULAR WEIGHT:2146.6STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [80448-90-4], RESEARCH AREA: PainREFERENCES: A. Goldstein et al., PNAS, 78, 7219 (1981); S. Tachibana et al., Nature, 295, 339 (1982); A.I. Faden, J. Neurosci., 12, 425 (1992)
Catalog No. 50-210-9207