Learn More
CPC Scientific H-Tyr-Gly-Gly-Phe-Leu-Arg-Arg-Ile-Arg-Pro-Lys-Leu-Lys-OH (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific DYNP007A
Encompass
Endogenous kappa-opioid receptor agonist that attenuates galanin-induced impairment of memory processes through the mediation of kappa-opioid receptors.ONE-LETTER SEQUENCE: YGGFLRRIRPKLKMOLECULAR FORMULA: C75H126N24O15MOLECULAR WEIGHT:1604STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [72957-38-1], RESEARCH AREA: PainREFERENCES: A. Goldstein et al., PNAS, 76, 6666 (1979)
Catalog No. 50-210-9213