Learn More
CPC Scientific H-Gly-Phe-Leu-Arg-Arg-Ile-Arg-Pro-Lys-Leu-Lys-Trp-Asp-Asn-Gln-OH (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific DYNP003B
Encompass
ONE-LETTER SEQUENCE: GFLRRIRPKLKWDNQMOLECULAR FORMULA: C88H143N29O20MOLECULAR WEIGHT:1927.3STORAGE CONDITIONS: -20 5CRESEARCH AREA: Pain
Catalog No. 50-210-9206