Learn More
Cayman Chemical DynorphIn AtrIfluoROCTAte 10mg
SDP
Catalog No. 502967324
Encompass_Preferred
A heptadecapeptide that acts as an endogenous opioid receptor agonist (Kis = 0.5-1 nM for κ, µ, and δ); has been implicated in antinociceptive functions
Catalog No. 50-296-7324 Supplier Cayman Chemical Supplier No. 1816910 MG
A heptadecapeptide that acts as an endogenous opioid receptor agonist (Kis = 0.5-1 nM for κ, µ, and δ); has been implicated in antinociceptive functions