Learn More
Cayman Chemical Dynorphin B trifluoroacetate
SDP
Supplier:  Cayman Chemical 18178500 UG
Encompass_Preferred
A 13-residue opioid peptide that acts as an endogenous κ1b-opioid receptor agonist (Ki = 1.1 nM); has been implicated in antinociceptive functions and has also been used to prime cardiogenesis in pluripotent embryonic stem cells
Catalog No. 50-296-6061