Learn More
Apexbio Technology LLC Dyphylline 479-18-5 50mg
SDP
Catalog No. 502240676
Encompass_Preferred
Dyphylline (CAS 479-18-5) is a synthetic xanthine derivative structurally related to theophylline It functions primarily as a phosphodiesterase inhibitor resulting in increased intracellular cAMP levels and subsequent smooth muscle relaxation Through these actions dyphylline exhibits bronchodilatory and vasodilatory properties making it useful in research investigating airway smooth muscle physiology and cardiovascular function Its pharmacological profile is of interest in preclinical studies focused on respiratory disorders such as asthma and chronic obstructive pulmonary disease as well as in the exploration of xanthine-mediated signaling pathways
This item is not returnable. View return policy
Catalog No. 50-224-0676 Supplier Apexbio Technology LLC Supplier No. A434350
Dyphylline (CAS 479-18-5) is a synthetic xanthine derivative structurally related to theophylline It functions primarily as a phosphodiesterase inhibitor resulting in increased intracellular cAMP levels and subsequent smooth muscle relaxation Through these actions dyphylline exhibits bronchodilatory and vasodilatory properties making it useful in research investigating airway smooth muscle physiology and cardiovascular function Its pharmacological profile is of interest in preclinical studies focused on respiratory disorders such as asthma and chronic obstructive pulmonary disease as well as in the exploration of xanthine-mediated signaling pathways