Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dypnone, Thermo Scientific Chemicals

Catalog No. AC465850050
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC465850050 5 g
AC465850250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC465850050 Supplier Thermo Scientific Chemicals Supplier No. 465850050
Add to Cart

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 495-45-4
Molecular Formula C16H14O
Molecular Weight (g/mol) 222.29
MDL Number MFCD00043705
InChI Key PLELHVCQAULGBH-OUKQBFOZSA-N
Synonym beta-Methylchalcone; beta-Methylstyryl phenyl ketone
IUPAC Name (2E)-1,3-diphenylbut-2-en-1-one
SMILES C\C(=C/C(=O)C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Alpha Vector DYPNONE
Color Colorless to Yellow
Infrared Spectrum Conforms
Packaging glass bottle
Quantity 5 g
Solubility Information insoluble aq. soln.
Formula Weight 222.29
Physical Form Liquid
Chemical Name or Material Dypnone
ShelfLife 5 years
storageNote1 normal conditions

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.