Learn More
Enzo Life Sciences DYRK kinase substrate (1mg)
SDP
Supplier:  Enzo Life Sciences BMLP2610001
Encompass_Preferred
This peptide (also known as Woodtide), KKISGRLSPIMTEQ, is derived from transcription factor FKHR with the addition of two lysines to facilitate binding to phosphocellulose membranes. It is a substrate for the DYRK family of kinases. Alternative name: Woodtide. Purity: ≥95% (HPLC). Formulation: Lyophilized. Solubility: Soluble in water. Long Term Storage: -20°C.
Catalog No. 50-201-1319