Learn More
AdipoGen DYRK1 Inhibitor Negative Control AnnH79 (500 µg)
SDP
Supplier:  AdipoGen AGCR13652C500
Encompass_Preferred
Chemical. Formula: C13H6BrCl2N3. MW: 355.0. Synthetic. Inactive harmine analog that does not inhibit DYRK1A/DYRK1B. Useful as negative control for β-carboline analog DYRK1A/B inhibitors.
Catalog No. 50-205-4027