Learn More
Carnabio Usa Inc DYRK1A/5UG/FULL-LENGTH
Supplier:  Carnabio Usa Inc 041305
Encompass
DYRK1A, Full-length, Wild type, Amino Acid 1-763, NP_001387.2, Expressed in Insect (sf21); 5microg
Catalog No. 50-754-0400