Learn More
Aviva Systems Biology Corp Dyrk1a Antibody - N-terminal region (ARP45796_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP45796_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Dyrk1a. It was validated on Western Blot. The immunogen is a synthetic peptide corresponding to a region of Mouse. Alternate names: 2310043O08Rik, D16Ertd272e, D16Ertd493e, Dyrk, ENSMUSG00000074897, Gm10783, MGC150253, MGC150254, Mnbh, Mp86, mmbApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-285-217