Learn More
Carnabio Usa Inc DYRK1B/5UG/FULL-LENGTH
Supplier:  Carnabio Usa Inc 041315
Encompass
DYRK1B, Full-length, Wild type, Amino Acid 1-629, NP_004705.1, Expressed in Insect (sf21); 5microg
Catalog No. 50-754-0401