Learn More
Aviva Systems Biology Corp DYRK2 Antibody - C-terminal region (ARP64303_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP64303_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against DYRK2. It was validated on Western Blot. . Alternate names: FLJ21217, FLJ21365ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-288-231