Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents QP10058BA20UG
Encompass_Preferred
Product Type: Recombinant Protein. Conjugate/Tag: His. Host: Baculovirus. Species Reactivity: Human. Protein Expression: A DNA sequence encoding the Homo sapiens (Human) DYRK2, Isoform 2, was expressed in the hosts and tags indicated. AA Sequence: See company website. Buffer: Tris-based buffer, 50% glycerol. Purity: Greater than 90% as determined by SDS-PAGE. Endotoxin: Not determined. Gene Symbol: DYRK2. Uniprot: Q92630-2.
Catalog No. 50-188-5485