Learn More
Sino Biological Recombinant Human Dmpk Protein (Full Length, GST Tag), Active
Catalog No. 502575415
Encompass_Preferred
Recombinant full length human DMPK was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
Catalog No. 50-257-5415 Supplier Sino Biological Supplier No. D0710G50
Recombinant full length human DMPK was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.