Learn More
Aviva Systems Biology Corp DYRK3 Antibody - N-terminal region (ARP30647_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP30647_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against DYRK3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human DYRK3. Alternate names: DYRK5, RED, REDK, hYAK3-2ApplicationWB. Species reactivity: Guinea Pig, Human, Rat, Yeast
Catalog No. 50-014-62877