Learn More
Aviva Systems Biology Corp DYSF Antibody - N-terminal region (ARP45477_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP45477_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against DYSF. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human DYSF. Alternate names: FER1L1, FLJ00175, FLJ90168, LGMD2B, MMD1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-59036