Learn More
Aviva Systems Biology Corp DYSFIP1 Antibody - middle region (ARP51794_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP51794_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against DYSFIP1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human DYSFIP1. Alternate names: MGC138299, DYSFIP1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat
Catalog No. 50-014-60616