Learn More
Sigma Organic Chemistry Dysprosium(III) acetate hydrate | 25G | 15280-55-4 | | 99.9% trace metals basis
Supplier: Sigma Organic Chemistry 32553825G

Dysprosium(III) acetate hydrate, 25G
About This Item:
Linear Formula: (CH3CO2)3Dy -+ 4H2O
UNSPSC Code: 12352300
Storage: room temp
EINECS Number: 242-565-8
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.