Learn More
Strem, An Ascensus Company CAS# 10025-74-8. 10g. Dysprosium(III) chloride, anhydrous (99.9%-Dy) (REO). MFCD00010984
SDP
Catalog No. 5090116111
Encompass
CAS# 10025-74-8. 10g. Dysprosium(III) chloride, anhydrous (99.9%-Dy) (REO). MFCD00010984. Molecular Weight: 268.86. Molecular Formula: DyCl3. Color/form: white pwdr. Strem# 93-6607. http://www.strem.com/catalog/v/93-6607/
Catalog No. 50-901-16111 Supplier Strem, An Ascensus Company Supplier No. 93660710G
CAS# 10025-74-8. 10g. Dysprosium(III) chloride, anhydrous (99.9%-Dy) (REO). MFCD00010984. Molecular Weight: 268.86. Molecular Formula: DyCl3. Color/form: white pwdr. Strem# 93-6607. http://www.strem.com/catalog/v/93-6607/