Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dysprosium(III) iodide, ultra dry, 99.99% (REO)

Catalog No. AA3569306
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3569306 5 g
AA3569303 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA3569306 Supplier Thermo Scientific Chemicals Supplier No. 03569306
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 15474-63-2 | DyI3 | 543.213 g/mol

It is used in high-intensity metal-halide lamps.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15474-63-2
Molecular Formula DyI3
Molecular Weight (g/mol) 543.213
MDL Number MFCD00016072
InChI Key RZQFCZYXPRKMTP-UHFFFAOYSA-K
Synonym dysprosium iii iodide, dysprosium triiodide, dysprosium iodide, acmc-20al3a, dysprosium iii ; iodide, dysprosium 3+ triiodide, dysprosium iii iodide, anhydrous
PubChem CID 84931
IUPAC Name dysprosium(3+);triiodide
SMILES [I-].[I-].[I-].[Dy+3]

Specifications

Melting Point 955°C
Color Green to Yellow
Physical Form Flakes
Quantity 5 g
Packaging Ampule
Sensitivity Hygroscopic
Solubility Information Soluble in water.
Formula Weight 543.21
Percent Purity 99.99%
Assay (REO)
Chemical Name or Material Dysprosium(III) iodide, Ultra dry
Show More Show Less
Hazard Category H317-H334
Hazard Statement GHS H Statement
H334-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause an allergic skin reaction.
Precautionary Statement P261-P272-P280g-P285-P302+P352-P304+P341-P333+P313-P342+P311-P363-P501c
EINECSNumber 239-493-4
TSCA Yes
storageNote1
  • Ampouled under argon
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.