Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dysprosium(III) nitrate pentahydrate, 99.9% (REO)

Catalog No. AA1292214
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1292214 25 g
AA1292222 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA1292214 Supplier Thermo Scientific Chemicals Supplier No. 01292214
  • $88.40
    $64.65 / Each of 1

    Save $23.75

    27% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 10031-49-9 | DyH10N3O14 | 438.59 g/mol

Dysprosium(III) nitrate pentahydrate is used to prepare carboxylate-based dinuclear dysprosium compound by reacting with n-butyric acid and triethylamine using methanol as a solvent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10031-49-9
Molecular Formula DyH10N3O14
Molecular Weight (g/mol) 438.59
MDL Number MFCD00149683
InChI Key NOTQUFQJAWMLCE-UHFFFAOYSA-N
IUPAC Name dysprosium(3+) pentahydrate trinitrate
SMILES O.O.O.O.O.[Dy+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Melting Point 88.6°C
Color Yellow
Physical Form Crystalline
Quantity 25 g
Assay Percent Range 99.9% (REO)
Solubility Information Soluble in water.
Formula Weight 438.59 (348.51 Anhydrous)
Sensitivity Hygroscopic
Chemical Name or Material Dysprosium(III) nitrate pentahydrate, Rare Earth Oxides Content
Hazard Category H272-H315-H319-H335
Hazard Statement Keep away from combustible material. In case of contact with eyes, rinse immediately with plenty of water and seek medical advice.
Precautionary Statement P210-P220-P221-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
DOTInformation DOT Class: 5.1, Packing Group: III
EINECSNumber 233-410-5
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.