Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dysprosium(III) nitrate pentahydrate, REacton™, 99.99% (REO)

Catalog No. AA1131514
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1131514 25 g
AA1131522 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA1131514 Supplier Thermo Scientific Chemicals Supplier No. 01131514
Add to Cart

CAS: 10031-49-9 | DyH10N3O14 | 438.59 g/mol

Dysprosium(III) nitrate is used for catalysts. It is used in the preparation of dysprosium oxide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10031-49-9
Molecular Formula DyH10N3O14
Molecular Weight (g/mol) 438.59
MDL Number MFCD00149683
InChI Key NOTQUFQJAWMLCE-UHFFFAOYSA-N
IUPAC Name dysprosium(3+) pentahydrate trinitrate
SMILES O.O.O.O.O.[Dy+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Melting Point 88.6°C
Color Cream to Yellow
pH 3
Physical Form Crystalline Aggregates
Quantity 25 g
UN Number UN1477
Merck Index 14,3482
Solubility Information Soluble in water.
Formula Weight 438.59 (348.51 Anhydrous)
Percent Purity 99.99%
Grade Reacton
Odor Odorless
Sensitivity Hygroscopic
Assay (REO)
Chemical Name or Material Dysprosium(III) nitrate pentahydrate
Show More Show Less
Hazard Category H272-H315-H319-H335
Hazard Statement GHS H Statement
H272-H315-H319-H335
May intensify fire
oxidizer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P220-P221-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
DOTInformation Transport Hazard Class: 5.1; Packing Group: III; Proper Shipping Name: NITRATES, INORGANIC, N.O.S.
EINECSNumber 233-410-5
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.