Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dysprosium(III) oxide, 99.9% (REO)

Catalog No. AA3637022
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3637022 100 g
AA3637014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA3637022 Supplier Thermo Scientific Chemicals Supplier No. 03637022
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 1308-87-8 | Dy2O3 | 373.00 g/mol

Dysprosium(III) oxide is uses in ceramics, glass, phosphors, lasers and dysprosium metal halide lamps.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1308-87-8
Molecular Formula Dy2O3
Molecular Weight (g/mol) 373.00
MDL Number MFCD00010986
InChI Key GEZAXHSNIQTPMM-UHFFFAOYSA-N
IUPAC Name didysprosium(3+) trioxidandiide
SMILES [O--].[O--].[O--].[Dy+3].[Dy+3]

Specifications

Melting Point 2340°C
Physical Form Sintered Lump
Quantity 100 g
Sensitivity Hygroscopic
Density 7.81 g/mL at 27°C
Merck Index 14,3482
Solubility Information Insoluble in water.
Formula Weight 373
Percent Purity 99.9%
Assay (REO)
Chemical Name or Material Dysprosium(III) oxide
Show More Show Less
EINECSNumber 215-164-0
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.