Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Dysprosium(III) Trifluoromethanesulfonate

Catalog No. 4256645G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
42-566-45G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Warning
Catalog No. 42-566-45G Supplier Sigma Aldrich Supplier No. 4256645G
  • NEW LOWER PRICE
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Dysprosium(III) trifluoromethanesulfonate (Dysprosium(III) triflate) is a lanthanide triflate.Dysprosium(III) trifluoromethanesulfonate, a water-tolerant Lewis acid, has been used in the following studies:Aldol reaction of silyl enol ethers with aldehydes. As an effective catalyst for electrophilic substitution reactions of indoles with imines.As catalyst for the synthesis of 4-aminocyclopentenones and functionalized azaspirocycles, via intramolecular aza-Piancatelli rearrangement.As new curing initiator to study the curing of diglycidyl ether of bisphenol-A (DGEBA).

Specifications

CAS 7175-81-7
Density 0.98 g/mL at 25°C
Boiling Point 55°C (18 mmHg, lit.)
Molecular Formula C5H11NO
Linear Formula C5H11NO
Quantity 5 g
MDL Number MFCD00085303
Synonym Dysprosium(III) triflate; Tris(triflato)dysprosium
Molecular Weight (g/mol) 101.15 g/mol
Biological Activity Respiratory System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.