Learn More
Bioss -Dystroglycan - Tyr-892 Phos
SDP
Catalog No. 5000065303
Encompass_Preferred
-Dystroglycan (Tyr-892) Primary Antibody Unconjugated Host Species Mouse Target Species Human Mouse Rat Clonality Monoclonal Applications WB M117Concentration Size 100 ul Storage Buffer PBS 1 mg/ml BSA 0 05% NaN3 and 50% glycerol
Catalog No. 50-000-65303 Supplier Bioss Supplier No. BSM70456M
-Dystroglycan (Tyr-892) Primary Antibody Unconjugated Host Species Mouse Target Species Human Mouse Rat Clonality Monoclonal Applications WB M117Concentration Size 100 ul Storage Buffer PBS 1 mg/ml BSA 0 05% NaN3 and 50% glycerol