Learn More
Aviva Systems Biology Corp DNAAF4 Antibody - C-terminal region (ARP52547_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP52547_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against DYX1C1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human DYX1C1. Alternate names: DYX1, DYXC1, EKN1, FLJ37882, MGC70618, RDApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-60769