Learn More
TARGETMOL CHEMICALS INC DZ2002 5MG
SDP
Catalog No. 502581566
Encompass_Preferred
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. DZ2002 is an orally active reversible and low-cytotoxic type III SAHH inhibitor (Ki=17.9 nM) with good immunosuppressive activity. DZ2002 prevents the development of experimental dermal fibrosis by reversing the profibrotic phenotype of various cell types. DZ2002 can be used in studies of autoimmune diseases such as lupus syndrome and systemic sclerosis. purity: 98%
Catalog No. 50-258-1566 Supplier TARGETMOL CHEMICALS INC Supplier No. T111345MG
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. DZ2002 is an orally active reversible and low-cytotoxic type III SAHH inhibitor (Ki=17.9 nM) with good immunosuppressive activity. DZ2002 prevents the development of experimental dermal fibrosis by reversing the profibrotic phenotype of various cell types. DZ2002 can be used in studies of autoimmune diseases such as lupus syndrome and systemic sclerosis. purity: 98%