Learn More
Apexbio Technology LLC 3-Deazaneplanocin A (DZNep) hydrochloride 120964-45-6 10mM (in 1mL DMSO)
SDP
Catalog No. 5000061577
Encompass_Preferred
3-Deazaneplanocin A (DZNep) hydrochloride is a selective inhibitor targeting histone methyltransferase EZH2 exhibiting inhibitory activity with an IC50 range of 0 08-0 24 M Additionally DZNep inhibits S-adenosylhomocysteine hydrolase (SAHH) at a Ki of 50 pM EZH2 acts as a catalytic subunit of polycomb repressive complex 2 (PRC2) participating in modulation of gene expression via histone methylation Research demonstrates that DZNep suppresses EZH2-dependent gene regulation pathways in cancer cells leading to altered expression of cell-cycle and apoptosis-associated genes DZNep is commonly utilized in epigenetic studies of various tumors and metabolic dysfunction models including AML liver fibrosis and lipid accumulation studies
Catalog No. 50-000-61577 Supplier Apexbio Technology LLC Supplier No. A81825.1
3-Deazaneplanocin A (DZNep) hydrochloride is a selective inhibitor targeting histone methyltransferase EZH2 exhibiting inhibitory activity with an IC50 range of 0 08-0 24 M Additionally DZNep inhibits S-adenosylhomocysteine hydrolase (SAHH) at a Ki of 50 pM EZH2 acts as a catalytic subunit of polycomb repressive complex 2 (PRC2) participating in modulation of gene expression via histone methylation Research demonstrates that DZNep suppresses EZH2-dependent gene regulation pathways in cancer cells leading to altered expression of cell-cycle and apoptosis-associated genes DZNep is commonly utilized in epigenetic studies of various tumors and metabolic dysfunction models including AML liver fibrosis and lipid accumulation studies