Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (E)-1-[(2R)-2-(2-hydroxyethyl)piperidin-1-yl]-3-(2-phenylpyrazolo[1,5-a]pyridin-3-yl)prop-2-en-1-one | 121524-18-3 | 99.2% | 375.46 g/mol | C23H25N3O2 | 1 MG
SDP

Catalog No. 5000243688
Encompass_Preferred

FK 453 is a research-grade small molecule adenosine A1 receptor antagonist with low-nanomolar potency (IC50 = 17.2 nM). It is supplied as a high-purity powder intended for in vitro pharmacology, receptor profiling, and other laboratory research applications.

  • Potent and selective adenosine A1 receptor antagonist.
  • Low-nanomolar potency (IC50 17.2 nM).
  • High purity (99.2%).
  • Molecular formula C23H25N3O2; molecular weight 375.46 g/mol.
  • Supplied as a powder with DMSO solubility for assay preparation.
  • Available in small-scale quantities suitable for research use.

Catalog No. 50-002-43688 Supplier Medchemexpress LLC Supplier No. HY105275A1MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

FK 453 is a research-grade small molecule adenosine A1 receptor antagonist with low-nanomolar potency (IC50 = 17.2 nM). It is supplied as a high-purity powder intended for in vitro pharmacology, receptor profiling, and other laboratory research applications.

  • Potent and selective adenosine A1 receptor antagonist.
  • Low-nanomolar potency (IC50 17.2 nM).
  • High purity (99.2%).
  • Molecular formula C23H25N3O2; molecular weight 375.46 g/mol.
  • Supplied as a powder with DMSO solubility for assay preparation.
  • Available in small-scale quantities suitable for research use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.