Learn More
Medchemexpress LLC (E)-1,3-Di(thiophen-2-yl)prop-2-en-1-one | 42811-88-1 | 99.4% | 220.31 | 250 MG
SDP
Catalog No. 5000402938
Encompass_Preferred
(E)-1,3-Di(thiophen-2-yl)prop-2-en-1-one is a chemical compound primarily used as a drug intermediate for the synthesis of various active compounds. It is intended for laboratory research by experienced personnel in appropriately equipped and authorized facilities and has not been fully validated for medical applications.
  • Used as a drug intermediate for synthesizing active compounds.
  • Intended for laboratory research.
  • Appears as a light yellow to yellow solid.
  • Requires storage in a cool, well-ventilated area, away from direct sunlight and ignition sources.
  • Store powder at -20°C for 3 years, or 4°C for 2 years.
  • When in solvent, store at -80°C for 6 months, or -20°C for 1 month.
  • Avoid breathing dust or vapors.
  • Wear appropriate personal protective equipment during handling.
Catalog No. 50-004-02938 Supplier Medchemexpress LLC Supplier No. 250MGHY43405
(E)-1,3-Di(thiophen-2-yl)prop-2-en-1-one is a chemical compound primarily used as a drug intermediate for the synthesis of various active compounds. It is intended for laboratory research by experienced personnel in appropriately equipped and authorized facilities and has not been fully validated for medical applications.
  • Used as a drug intermediate for synthesizing active compounds.
  • Intended for laboratory research.
  • Appears as a light yellow to yellow solid.
  • Requires storage in a cool, well-ventilated area, away from direct sunlight and ignition sources.
  • Store powder at -20°C for 3 years, or 4°C for 2 years.
  • When in solvent, store at -80°C for 6 months, or -20°C for 1 month.
  • Avoid breathing dust or vapors.
  • Wear appropriate personal protective equipment during handling.