Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (E)-1-(4-acetylpiperazin-1-yl)-3-[3-nitro-4-(2-propan-2-ylphenyl)sulfanylphenyl]prop-2-en-1-one | 280749-17-9 | 99.3% | 453.55 g·mol⁻¹ | C24H27N3O4S | 1 ML
SDP

Catalog No. 5000241612
Encompass_Preferred

A small-molecule allosteric inhibitor of the LFA-1/ICAM-1 interaction used as a research tool to study LFA-1-mediated cell adhesion and signaling. Reported activity includes IC50 = 44 nM in an LFA-1/ICAM-1 binding assay and IC50 = 35 nM in a cellular adhesion assay. Supplied as a 10 mM solution in DMSO (1 mL) and as various solid pack sizes. Intended for research use only.

  • Potent allosteric inhibitor of LFA-1/ICAM-1 interaction.
  • Reported IC50: 44 nM (binding) and 35 nM (cell adhesion).
  • Available as 10 mM solution in DMSO and multiple solid sizes.
  • High purity (99.33%).
  • Molecular weight 453.55 g·mol⁻¹; formula C24H27N3O4S.
  • Light yellow to green yellow solid appearance.
  • For research use only; not for human or clinical use.

Catalog No. 50-002-41612 Supplier Medchemexpress LLC Supplier No. HY10758710MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

A small-molecule allosteric inhibitor of the LFA-1/ICAM-1 interaction used as a research tool to study LFA-1-mediated cell adhesion and signaling. Reported activity includes IC50 = 44 nM in an LFA-1/ICAM-1 binding assay and IC50 = 35 nM in a cellular adhesion assay. Supplied as a 10 mM solution in DMSO (1 mL) and as various solid pack sizes. Intended for research use only.

  • Potent allosteric inhibitor of LFA-1/ICAM-1 interaction.
  • Reported IC50: 44 nM (binding) and 35 nM (cell adhesion).
  • Available as 10 mM solution in DMSO and multiple solid sizes.
  • High purity (99.33%).
  • Molecular weight 453.55 g·mol⁻¹; formula C24H27N3O4S.
  • Light yellow to green yellow solid appearance.
  • For research use only; not for human or clinical use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.