Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(E)-1-Benzyloxy-2-methoxy-4-(1-propenyl)benzene 98.0+%, TCI America™
SDP

Catalog No. B144125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B144125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B144125G Supplier TCI America Supplier No. B144125G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 92666-21-2
Molecular Formula C17H18O2
Molecular Weight (g/mol) 254.33
MDL Number MFCD00026985
InChI Key YKSSSKBJDZDZTD-UHFFFAOYSA-N
Synonym trans-Benzylisoeugenol
PubChem CID 764139
IUPAC Name 1-(benzyloxy)-2-methoxy-4-(prop-1-en-1-yl)benzene
SMILES COC1=C(OCC2=CC=CC=C2)C=CC(C=CC)=C1

Specifications

Melting Point 59°C
Color White
Quantity 25 g
Formula Weight 254.33
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material (E)-1-Benzyloxy-2-methoxy-4-(1-propenyl)benzene
EINECSNumber (3)-0634
RTECSNumber CY8225000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.