Learn More
Medchemexpress LLC HY-U00050 5mg Medchemexpress, (E)-10-Hydroxynortriptyline CAS:47132-16-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYU000505MG
Encompass_Preferred
Medchemexpress, HY-U00050 5mg (E)-10-Hydroxynortriptyline CAS:47132-16-1 (E)-10-Hydroxynortriptyline (E-10-OH-NT) is a metabolite of Nortriptyline (HY-B1417). Nortriptyline is a tricyclic antidepressant and the main active metabolite of Amitriptyline (HY-B0527A). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-3184