Learn More
Medchemexpress LLC (E)-10-Hydroxynortriptyline (E-10-OH-NT) | 47132-16-1 | 279.38 | 1 MG
SDP
Catalog No. 5000446542
Encompass_Preferred
(E)-10-Hydroxynortriptyline (E-10-OH-NT) is a metabolite of Nortriptyline, a tricyclic antidepressant and the primary active metabolite of Amitriptyline. This compound is approximately 50% as potent as nortriptyline as an inhibitor of neuronal norepinephrine uptake in vitro and exhibits fewer anticholinergic effects in humans. It presents as a white to off-white solid with the molecular formula C19H21NO.
  • Metabolite of Nortriptyline
  • Inhibits neuronal norepinephrine uptake
  • Demonstrates fewer anticholinergic effects
  • Appears as a white to off-white solid
  • Intended for research use only
Catalog No. 50-004-46542 Supplier Medchemexpress LLC Supplier No. HYU000501MG
(E)-10-Hydroxynortriptyline (E-10-OH-NT) is a metabolite of Nortriptyline, a tricyclic antidepressant and the primary active metabolite of Amitriptyline. This compound is approximately 50% as potent as nortriptyline as an inhibitor of neuronal norepinephrine uptake in vitro and exhibits fewer anticholinergic effects in humans. It presents as a white to off-white solid with the molecular formula C19H21NO.
  • Metabolite of Nortriptyline
  • Inhibits neuronal norepinephrine uptake
  • Demonstrates fewer anticholinergic effects
  • Appears as a white to off-white solid
  • Intended for research use only