Learn More
Miele, Inc. E 103/1 Quarter basket insert
Catalog No. 502870781
Encompass_Preferred
E 103/1 Quarter basket insert for up to 200 test tubes 12x75mm, includes A13
Catalog No. 50-287-0781 Supplier Miele, Inc. Supplier No. 12667340
E 103/1 Quarter basket insert for up to 200 test tubes 12x75mm, includes A13