Learn More
Miele, Inc. E 103/1 Quarter Insert (160 test tubes, 75 mm maximum length)
Catalog No. 501713015
Encompass_Preferred
E 103/1 Mesh Basket
For the optimum loading of test tubes up to 12 x 75 mm.
Suitable for about 200 Test tubes (up to 12 x 75 mm)
Divided into 6 sections
The mesh width is 8 x 8 mm
Including lid A 13
Visit www.mieleusa.com/professional for technical information.
Catalog No. 50-171-3015 Supplier Miele, Inc. Supplier No. 06907630
E 103/1 Mesh Basket
For the optimum loading of test tubes up to 12 x 75 mm.
Suitable for about 200 Test tubes (up to 12 x 75 mm)
Divided into 6 sections
The mesh width is 8 x 8 mm
Including lid A 13
Visit www.mieleusa.com/professional for technical information.