Learn More
Miele, Inc. E 104/1 Quarter basket insert
Catalog No. 502870720
Encompass_Preferred
E 104/1 Quarter basket insert for up to 200 test tubes 12x105mm, includes A13
Catalog No. 50-287-0720 Supplier Miele, Inc. Supplier No. 12667370
E 104/1 Quarter basket insert for up to 200 test tubes 12x105mm, includes A13