Learn More
Miele, Inc. E 104/1 Quarter Insert (160 test tubes, 105 mm maximum length)
Catalog No. 501713016
Encompass_Preferred
E 104/1 Mesh Basket
For the optimum loading of test tubes up to 12 x 105 mm.
Suitable for about 200 Test tubes (up to 12 x 105 mm)
Capacity including lid A 13
The mesh width is 8 x 8 mm
Visit www.mieleusa.com/professional for technical information.
Catalog No. 50-171-3016 Supplier Miele, Inc. Supplier No. 06907640
Add to Cart
Add to Cart
E 104/1 Mesh Basket
For the optimum loading of test tubes up to 12 x 105 mm.
Suitable for about 200 Test tubes (up to 12 x 105 mm)
Capacity including lid A 13
The mesh width is 8 x 8 mm
Visit www.mieleusa.com/professional for technical information.