Learn More
Miele, Inc. E 105/1 Quarter basket insert
Catalog No. 502870698
Encompass_Preferred
E 105/1 Quarter basket insert for up to 200 test tubes 12x165mm, includes A13
Catalog No. 50-287-0698 Supplier Miele, Inc. Supplier No. 12667380
E 105/1 Quarter basket insert for up to 200 test tubes 12x165mm, includes A13