Learn More
Miele, Inc. E 105/1 Quarter Insert (160 test tubes, 165 mm maximum length)
Catalog No. 501713017
Encompass_Preferred
E 105/1 Mesh Basket
For the optimum loading of test tubes up to 12 x 165 mm.
Suitable for about 200 Test tubes (up to 12 x 165 mm)
Capacity including lid A 13
The mesh width is 9 x 9 mm
Visit www.mieleusa.com/professional for technical information.
Catalog No. 50-171-3017 Supplier Miele, Inc. Supplier No. 06907650
Add to Cart
Add to Cart
E 105/1 Mesh Basket
For the optimum loading of test tubes up to 12 x 165 mm.
Suitable for about 200 Test tubes (up to 12 x 165 mm)
Capacity including lid A 13
The mesh width is 9 x 9 mm
Visit www.mieleusa.com/professional for technical information.