Learn More
Miele, Inc. E 106/2 Half Insert with spring clips (15 clips 175 mm tall)
Catalog No. 501713013
Encompass_Preferred
E 106/2 Mesh Basket
For the optimum loading of wide neck flasks, measuring cylinders, etc.
Equipped with 13 large spring hooks x 175 mm height
The distance between the spring hooks is approx. 85 mm
Visit www.mieleusa.com/professional for technical information.
Catalog No. 50-171-3013 Supplier Miele, Inc. Supplier No. 03808330
E 106/2 Mesh Basket
For the optimum loading of wide neck flasks, measuring cylinders, etc.
Equipped with 13 large spring hooks x 175 mm height
The distance between the spring hooks is approx. 85 mm
Visit www.mieleusa.com/professional for technical information.