Learn More
Miele, Inc. E 110 Half Insert for beakers (10 beakers, 250 - 600 ml)
Catalog No. NC0901666
Encompass_Preferred
E 110 Mesh Basket
For the optimum loading of beakers from 250 to 600 ml.
Suitable for 10 glass beakers between 250 - 600 ml
Equipped with 10 x 3 stay bars
For use in lower basket
Visit www.mieleusa.com/professional for technical information.
Catalog No. NC0901666 Supplier Miele, Inc. Supplier No. 03808390
E 110 Mesh Basket
For the optimum loading of beakers from 250 to 600 ml.
Suitable for 10 glass beakers between 250 - 600 ml
Equipped with 10 x 3 stay bars
For use in lower basket
Visit www.mieleusa.com/professional for technical information.