Learn More
Miele, Inc. E 111 Half Insert for beakers (8 beakers, 600 - 1000 ml)
Catalog No. 501713014
Encompass_Preferred
E 111 Mesh Basket
For the optimum loading of beakers from 600 to 1,000 ml.
Suitable for 8 glass beakers between 600 - 1000 ml
Equipped with 8 x 3 stay bars
For use in lower basket
Visit www.mieleusa.com/professional for technical information.
Catalog No. 50-171-3014 Supplier Miele, Inc. Supplier No. 03808420
E 111 Mesh Basket
For the optimum loading of beakers from 600 to 1,000 ml.
Suitable for 8 glass beakers between 600 - 1000 ml
Equipped with 8 x 3 stay bars
For use in lower basket
Visit www.mieleusa.com/professional for technical information.