Learn More
Miele, Inc. E 134 Half Insert for 210 slides
Catalog No. 501713020
Encompass_Preferred
E 134 Mesh Basket
For optimum loading of 210 slides.
Suitable for 210 slides
Equipped with 26 compartments at 26 x 11 mm, wire thickness 3 mm
Visit www.mieleusa.com/professional for technical information.
Catalog No. 50-171-3020 Supplier Miele, Inc. Supplier No. 03808600
E 134 Mesh Basket
For optimum loading of 210 slides.
Suitable for 210 slides
Equipped with 26 compartments at 26 x 11 mm, wire thickness 3 mm
Visit www.mieleusa.com/professional for technical information.