Learn More
Miele, Inc. E 139/1 Quarter basket insert
Catalog No. 502870802
Encompass_Preferred
E 139/1 Quarter basket insert for up to 200 test tubes 12x200mm, includes A13
Catalog No. 50-287-0802 Supplier Miele, Inc. Supplier No. 12667390
Add to Cart
Add to Cart
E 139/1 Quarter basket insert for up to 200 test tubes 12x200mm, includes A13