Learn More
Miele, Inc. E 139/1 Quarter Insert (160 test tubes, 200 mm maximum length)
Catalog No. 501713018
Encompass_Preferred
E 139/1 Mesh Basket
For the optimum loading of test tubes up to 12 x 200 mm.
Suitable for test tubes up to 12 x 200 mm
Divided into 6 sections
Mesh width 9 x 9 mm
For use in lower basket
Including lid A 13
Visit www.mieleusa.com/professional for technical information.
Catalog No. 50-171-3018 Supplier Miele, Inc. Supplier No. 06907660
E 139/1 Mesh Basket
For the optimum loading of test tubes up to 12 x 200 mm.
Suitable for test tubes up to 12 x 200 mm
Divided into 6 sections
Mesh width 9 x 9 mm
For use in lower basket
Including lid A 13
Visit www.mieleusa.com/professional for technical information.